Learn More
Medchemexpress LLC Cyclic GMP-AMP disodium | 2407516-83-8 | 98.5% | 718.37 g/mol | C20H22N10Na2O13P2 | 5 MG
SDP
Catalog No. 5000263285
Encompass_Preferred
cGAMP disodium is the disodium salt of cyclic GMP-AMP, a cyclic dinucleotide STING agonist used in research to activate innate immune signaling and induce type I interferon production. It is provided as a white to off-white solid with high reported purity and good aqueous solubility for use in cellular and biochemical assays.
  • High purity suitable for research applications.
  • Good water solubility; dissolves in water and PBS with ultrasonic assistance.
  • Supplied in small research pack sizes for convenient handling.
  • Stable when stored sealed and protected from moisture.
  • Used to activate STING-mediated innate immune pathways in vitro.
Catalog No. 50-002-63285 Supplier Medchemexpress LLC Supplier No. HY1103855MG
cGAMP disodium is the disodium salt of cyclic GMP-AMP, a cyclic dinucleotide STING agonist used in research to activate innate immune signaling and induce type I interferon production. It is provided as a white to off-white solid with high reported purity and good aqueous solubility for use in cellular and biochemical assays.
  • High purity suitable for research applications.
  • Good water solubility; dissolves in water and PBS with ultrasonic assistance.
  • Supplied in small research pack sizes for convenient handling.
  • Stable when stored sealed and protected from moisture.
  • Used to activate STING-mediated innate immune pathways in vitro.