Learn More
Medchemexpress LLC Cyclic GMP-AMP (disodium); 3',3'-cGAMP (disodium) | 2407516-83-8 | 98.47% | 718.37 | 1 MG
SDP
Catalog No. 5000365454
Encompass_Preferred
cGAMP disodium is an endogenous second messenger that triggers interferon production in response to cytosolic DNA. It activates stimulator of interferon genes (STING), leading to a signaling cascade that produces type I interferons and other immune mediators. This product is suitable for various research applications.
  • Promotes antigen-specific proliferation capacity of spleen cells
  • Directly activates murine and human dendritic cells in vitro
  • Increases transcription of IFNB1 in stimulated fibroblasts
  • Induces an antiviral state and type I IFN secretion by activating ER-resident receptor STING
Catalog No. 50-003-65454 Supplier Medchemexpress LLC Supplier No. 1MGHY110385
cGAMP disodium is an endogenous second messenger that triggers interferon production in response to cytosolic DNA. It activates stimulator of interferon genes (STING), leading to a signaling cascade that produces type I interferons and other immune mediators. This product is suitable for various research applications.
  • Promotes antigen-specific proliferation capacity of spleen cells
  • Directly activates murine and human dendritic cells in vitro
  • Increases transcription of IFNB1 in stimulated fibroblasts
  • Induces an antiviral state and type I IFN secretion by activating ER-resident receptor STING