Learn More
Enzo Life Sciences cGMP Complete ELISA kit (1x96wells)
SDP
Catalog No. 50658190
Encompass_Preferred
Most sensitive (0.043 pmol/ml) and complete colorimetric kit for intra- and extracellular cGMP quantification. Alternative Name: cyclic GMP. Sensitivity: Assay Buffer : 0.42 pmol/ml (non-acetylated) 0.043 pmol/ml (acetylated) HCl: 0.604 pmol/ml (non-acetylated) 0.059 pmol/ml (acetylated) (range 0.8 - 500 pmol/ml (non-acetylated); 0.08 - 50 pmol/ml (acetylated)). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, 0.1M HCl, Assay buffer 2, Neutralizing reagent, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Triethylamine, Acetic anhydride
Catalog No. 50-658-190 Supplier Enzo Life Sciences Supplier No. ADI900164
Most sensitive (0.043 pmol/ml) and complete colorimetric kit for intra- and extracellular cGMP quantification. Alternative Name: cyclic GMP. Sensitivity: Assay Buffer : 0.42 pmol/ml (non-acetylated) 0.043 pmol/ml (acetylated) HCl: 0.604 pmol/ml (non-acetylated) 0.059 pmol/ml (acetylated) (range 0.8 - 500 pmol/ml (non-acetylated); 0.08 - 50 pmol/ml (acetylated)). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, 0.1M HCl, Assay buffer 2, Neutralizing reagent, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Triethylamine, Acetic anhydride