Learn More
Enzo Life Sciences Direct cGMP ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901014
Encompass_Preferred
Most sensitive (0.025 pmol/ml) colorimetric kit for intracellular cGMP quantification. Alternative Name: cyclic GMP. Sensitivity: Non-acetylated format: 0.604 pmol/ml. Acetylated 2-hour format: 0.059 pmol/ml. Acetylated overnight format: 0.025 pmol/ml . Range: (non-acetylated): 0.8 - 500 pmol/ml. Range: (acetylated): 0.08 - 50 pmol/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR Microtiter plate, Conjugate, Antibody, 0.1M HCl, Neutralizing reagent, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine
Catalog No. 50-658-206