Learn More
Enzo Life Sciences cGMP ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900013
Encompass_Preferred
Most sensitive (0.088 pmol/ml) colorimetric kit for extracellular cGMP quantification. This is a colorimetric competitive immunoassay kit for the quantitative determination of extracellular cGMP. Sensitivity is increased by acetylation (reagents included). Absorbance is read at 405 nm.The ready-to-use liquid color-coded reagents provided reduce error. Alternative Name: cyclic GMP. Sensitivity: (non-acetylated) 0.37 pmol/ml; (acetylated) 0.088 pmol/ml (range (non-acetylated) 0.16 - 500 pmol/ml; (acetylated) 0.16 - 100 pmol/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer 2, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine
Catalog No. 50-658-100