Learn More
Medchemexpress LLC Cyclic GMP sodium | 40732-48-7 | 99.9% | 367.19 | 25 MG
SDP
Catalog No. 5000437632
Encompass_Preferred
Cyclic GMP (cGMP) sodium is an important second messenger and a major intracellular mediator of extracellular signals like nitric oxide (NO) and natriuretic peptides (NPs). Its effects are mediated through three main groups of cellular targets: cGMP-dependent protein kinases (PKGs), cGMP-gated cation channels, and phosphodiesterases (PDEs). It can inhibit both platelet adhesion and aggregation, and its conjugate, cGAMP, can enhance antiviral immune responses.
  • Important second messenger.
  • Major intracellular mediator of extracellular signals such as nitric oxide (NO) and natriuretic peptides (NPs).
  • Mediates effects through cGMP-dependent protein kinases, cGMP-gated cation channels, and PDEs.
  • Inhibits platelet adhesion and aggregation.
  • Can enhance antiviral immune responses through its conjugate.
  • Activates cGMP-stimulated phosphodiesterases synergistically with Isoprenaline.
Catalog No. 50-004-37632 Supplier Medchemexpress LLC Supplier No. 25MGHY113469A
Cyclic GMP (cGMP) sodium is an important second messenger and a major intracellular mediator of extracellular signals like nitric oxide (NO) and natriuretic peptides (NPs). Its effects are mediated through three main groups of cellular targets: cGMP-dependent protein kinases (PKGs), cGMP-gated cation channels, and phosphodiesterases (PDEs). It can inhibit both platelet adhesion and aggregation, and its conjugate, cGAMP, can enhance antiviral immune responses.
  • Important second messenger.
  • Major intracellular mediator of extracellular signals such as nitric oxide (NO) and natriuretic peptides (NPs).
  • Mediates effects through cGMP-dependent protein kinases, cGMP-gated cation channels, and PDEs.
  • Inhibits platelet adhesion and aggregation.
  • Can enhance antiviral immune responses through its conjugate.
  • Activates cGMP-stimulated phosphodiesterases synergistically with Isoprenaline.