Learn More
Medchemexpress LLC Cyclic GMP (TBAOH) | 99.1% | 604.68 | 1 ML
SDP
Catalog No. 5000437485
Encompass_Preferred
Cyclic GMP (cGMP) TBAOH is an important second messenger and a major intracellular mediator of extracellular signals such as nitric oxide (NO) and natriuretic peptides (NPs). Its effects occur through three main groups of cellular targets: cGMP-dependent protein kinases (PKGs), cGMP-gated cation channels, and PDEs. Cyclic GMP can inhibit both platelet adhesion and aggregation. cGAMP (Cyclic-GMP-AMP), a conjugate of Cyclic GMP and AMP, can induce IRF3 phosphorylation and nuclear translocation, enhancing antiviral immune responses.
  • Important second messenger
  • Major intracellular mediator of extracellular signals such as nitric oxide (NO) and natriuretic peptides (NPs)
  • Affects cGMP-dependent protein kinases (PKGs), cGMP-gated cation channels, and PDEs
  • Can inhibit both platelet adhesion and aggregation
  • A conjugate can enhance antiviral immune responses
Catalog No. 50-004-37485 Supplier Medchemexpress LLC Supplier No. HY113469B10MM/1ML
Cyclic GMP (cGMP) TBAOH is an important second messenger and a major intracellular mediator of extracellular signals such as nitric oxide (NO) and natriuretic peptides (NPs). Its effects occur through three main groups of cellular targets: cGMP-dependent protein kinases (PKGs), cGMP-gated cation channels, and PDEs. Cyclic GMP can inhibit both platelet adhesion and aggregation. cGAMP (Cyclic-GMP-AMP), a conjugate of Cyclic GMP and AMP, can induce IRF3 phosphorylation and nuclear translocation, enhancing antiviral immune responses.
  • Important second messenger
  • Major intracellular mediator of extracellular signals such as nitric oxide (NO) and natriuretic peptides (NPs)
  • Affects cGMP-dependent protein kinases (PKGs), cGMP-gated cation channels, and PDEs
  • Can inhibit both platelet adhesion and aggregation
  • A conjugate can enhance antiviral immune responses