Learn More
Cayman Chemical Cyclic GMP (sodium salt), Purity: ≥98%, CAS: 40732-48-7, 50 mg
SDP
Catalog No. 502966184
Encompass_Preferred
A second messenger that is biosynthesized from GTP by guanylate cyclases; activates protein kinase G (PKG) and modulates ion channel conductance, with signaling affecting diverse processes including smooth muscle relaxation and proliferation, phototransduction, and energy homeostasis
Catalog No. 50-296-6184 Supplier Cayman Chemical Supplier No. 1882150 MG
A second messenger that is biosynthesized from GTP by guanylate cyclases; activates protein kinase G (PKG) and modulates ion channel conductance, with signaling affecting diverse processes including smooth muscle relaxation and proliferation, phototransduction, and energy homeostasis