Learn More
Biomatik Corporation Cyclic Guanosine Monophosphate (cGMP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12142
Encompass_Preferred
Cyclic Guanosine Monophosphate (cGMP) Mini sample ELISA Kit for detecting cGMP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 123.5-10,000pg/ml. Sensitivity: 49.6pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: c-GMP; 3',5'-Cyclic GMP; Cyclic 3',5'-GMP; Guanosine 3',5'-Cyclic Phosphate. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7069