Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich cyclic Isopropylidene 1,1-cyclopropanedicarboxylate

Catalog No. 3412665G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-126-65G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-126-65G Supplier Sigma Aldrich Supplier No. 3412665G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

cyclic Isopropylidene 1,1-cyclopropanedicarboxylate

Specifications

CAS 5617-70-9
Melting Point 63°C to 65°C (lit.)
Molecular Formula C8H10O4
Linear Formula C8H10O4
MDL Number MFCD00042796
Quantity 100 mg
Synonym 1,1-Cyclopropanedicarboxylic acid cyclic isopropylidene ester; 6,6-Dimethyl-5,7-dioxaspiro[2.5 ]octane-4,8-dione
Molecular Weight (g/mol) 170.16
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.