Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclic N-acetyl-D-mannosamine | 7772-94-3 | ≥98.0% | 221.21 | C8H15NO6 | 5 G
SDP

Catalog No. 5000208660
Encompass_Preferred

Cyclic N-acetyl-D-mannosamine is an endogenous monosaccharide metabolite used as a reference standard for research into sialic acid biosynthesis and glycan metabolism. It is provided as a high-purity research chemical with available solid and solution formats and an accompanying certificate of analysis.

  • Endogenous monosaccharide metabolite
  • Purity ≥98.0% (NMR)
  • Molecular formula C8H15NO6
  • Molecular weight 221.21
  • Available as solid and solution-ready formats
  • Suitable for biochemical and cellular research
  • Supplied with certificate of analysis

Catalog No. 50-002-08660 Supplier Medchemexpress LLC Supplier No. HYW0401545G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclic N-acetyl-D-mannosamine is an endogenous monosaccharide metabolite used as a reference standard for research into sialic acid biosynthesis and glycan metabolism. It is provided as a high-purity research chemical with available solid and solution formats and an accompanying certificate of analysis.

  • Endogenous monosaccharide metabolite
  • Purity ≥98.0% (NMR)
  • Molecular formula C8H15NO6
  • Molecular weight 221.21
  • Available as solid and solution-ready formats
  • Suitable for biochemical and cellular research
  • Supplied with certificate of analysis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.