Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclic N-acetyl-D-mannosamine | 7772-94-3 | MFCD00046758 | >98.0% | 221.21 g/mol | C8H15NO6 | 1 ML
SDP

Catalog No. 5000209489
Encompass_Preferred

Cyclic N-acetyl-D-mannosamine is an endogenous monosaccharide metabolite supplied for research use in both solid and prepared-solution formats. Vendor documentation and batch analysis describe high purity, detailed solubility data for aqueous and DMSO solutions, and recommended storage conditions for powder and solvent stocks.

  • Endogenous monosaccharide metabolite used in biochemical research.
  • Purity reported ≥98.0% by NMR for the referenced batch (vendor listings also show a 95.0% variant).
  • Available as a 10 mM, 1 mL solution in DMSO and as solids in multiple sizes (100 mg-5 g; larger sizes by quote).
  • Highly soluble in water and DMSO (vendor-reported water solubility 100 mg/mL with ultrasonic and warming).
  • Provides vendor guidance for preparing stock solutions and in vivo formulations.
  • Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-09489 Supplier Medchemexpress LLC Supplier No. HYW04015410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclic N-acetyl-D-mannosamine is an endogenous monosaccharide metabolite supplied for research use in both solid and prepared-solution formats. Vendor documentation and batch analysis describe high purity, detailed solubility data for aqueous and DMSO solutions, and recommended storage conditions for powder and solvent stocks.

  • Endogenous monosaccharide metabolite used in biochemical research.
  • Purity reported ≥98.0% by NMR for the referenced batch (vendor listings also show a 95.0% variant).
  • Available as a 10 mM, 1 mL solution in DMSO and as solids in multiple sizes (100 mg-5 g; larger sizes by quote).
  • Highly soluble in water and DMSO (vendor-reported water solubility 100 mg/mL with ultrasonic and warming).
  • Provides vendor guidance for preparing stock solutions and in vivo formulations.
  • Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.