Learn More
Medchemexpress LLC Cyclic nona-L-arginine hydrochloride | 98.4% | 1442.13 | 10 MG
SDP
Catalog No. 5000397321
Encompass_Preferred
Cyclic nona-L-arginine hydrochloride is a nonaarginine peptide primarily utilized for drug delivery applications. This peptide is notable for its enhanced translocation capabilities compared to its linear counterparts, offering potential advantages in research involving targeted delivery systems.
  • Translocates faster than linear counterparts
  • Used for drug delivery applications
Catalog No. 50-003-97321 Supplier Medchemexpress LLC Supplier No. HYP3193A10MG
Cyclic nona-L-arginine hydrochloride is a nonaarginine peptide primarily utilized for drug delivery applications. This peptide is notable for its enhanced translocation capabilities compared to its linear counterparts, offering potential advantages in research involving targeted delivery systems.
  • Translocates faster than linear counterparts
  • Used for drug delivery applications