Learn More
Cayman Chemical Cyclic Pifithrin-a hydrobrom
SDP
Catalog No. 502971473
Encompass_Preferred
A stable analog of PFT-α, formed by the condensation of PFT-α in solution; inhibits the growth of the cancer cell lines IGROV-1, A2780, and HCT116 (IC50 = 23, 77, and 103 μM, respectively); induces autophagy in HCT116 cells and sensitizes IGROV-1 and H460 cells to anti-microtubule agents
Catalog No. 50-297-1473 Supplier Cayman Chemical Supplier No. 1474850 MG
A stable analog of PFT-α, formed by the condensation of PFT-α in solution; inhibits the growth of the cancer cell lines IGROV-1, A2780, and HCT116 (IC50 = 23, 77, and 103 μM, respectively); induces autophagy in HCT116 cells and sensitizes IGROV-1 and H460 cells to anti-microtubule agents