Learn More
STEMCELL Technologies Cyclic Pifithrin-Alpha (Hydrobromide), Size: 10 mg
SDP
Catalog No. 502390196
Encompass
Cyclic Pifthrin-Alpha is a cell-permeable and reversible inhibitor of p53-mediated apoptosis and p53-dependent gene transcription. It is a more stable and less cytotoxic analog of the non-cyclic form of pifithrin-alpha. Cyclic Pifithrin-Alpha has also been reported to activate the aryl hydrocarbon receptor. (Fernandez-Cruz et al. Gary Jensen Komarov et al.)
Catalog No. 50-239-0196 Supplier STEMCELL Technologies Supplier No. 72064
Cyclic Pifthrin-Alpha is a cell-permeable and reversible inhibitor of p53-mediated apoptosis and p53-dependent gene transcription. It is a more stable and less cytotoxic analog of the non-cyclic form of pifithrin-alpha. Cyclic Pifithrin-Alpha has also been reported to activate the aryl hydrocarbon receptor. (Fernandez-Cruz et al. Gary Jensen Komarov et al.)