Learn More
Abcam Cyclic Pifithrin-alpha hydrobromide, pifithrin-alpha analog, 5MG
Catalog No. 502705880
Encompass_Preferred
MW 349.3 Da, Purity >98%. Stable, cell-permeable analog of pifithrin-α (ab120478). Inhibits p53-mediated apoptosis and p53-dependent gene transcription. Prevents dexamethasone-induced cell death in murine thymocytes (EC₅₀ = 2.01 μM). Potent STAT6 transcriptional inhibitor. Active in vivo and in vitro. See similar compound .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5880 Supplier Abcam Supplier No. AB1443275MG
MW 349.3 Da, Purity >98%. Stable, cell-permeable analog of pifithrin-α (ab120478). Inhibits p53-mediated apoptosis and p53-dependent gene transcription. Prevents dexamethasone-induced cell death in murine thymocytes (EC₅₀ = 2.01 μM). Potent STAT6 transcriptional inhibitor. Active in vivo and in vitro. See similar compound .
The product is subject to the following: Abcam Restricted Use Statement