Learn More
Selleck Chemical LLC Cyclic Pifithrin- hydrobromide 10mg 511296-88-1
SDP
Supplier:  Selleck Chemical LLC S579110MG
Encompass_Preferred
Cyclic Pifithrin-? hydrobromide is the hydrobromide salt form of Cyclic Pifithrin-?, which is a transcriptional inhibitor of p53. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8309