Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclic psap peptide | 1810853-35-0 | 99.9% | 639.74 | 1 MG
SDP

Catalog No. 5000399416
Encompass_Preferred

Cyclic PSAP peptide is a cyclic pentapeptide (DWLPK) that exhibits agent-like properties. It can inhibit metastatic spread and restrain tumor development in general in vivo. This product is for research use only.

  • Purity: 99.88%
  • Solid, white to off-white form
  • Sequence: Cyclic(Asp-Trp-Leu-Pro-Lys) / Cyclic(DWLPK)
  • For research use only
  • Sealed storage, away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO: 50 mg/mL (78.16 mM), requires ultrasonic treatment

Catalog No. 50-003-99416 Supplier Medchemexpress LLC Supplier No. HYP41021MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cyclic PSAP peptide is a cyclic pentapeptide (DWLPK) that exhibits agent-like properties. It can inhibit metastatic spread and restrain tumor development in general in vivo. This product is for research use only.

  • Purity: 99.88%
  • Solid, white to off-white form
  • Sequence: Cyclic(Asp-Trp-Leu-Pro-Lys) / Cyclic(DWLPK)
  • For research use only
  • Sealed storage, away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO: 50 mg/mL (78.16 mM), requires ultrasonic treatment

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.