Learn More
Medchemexpress LLC Cyclic PSAP peptide | 1810853-35-0 | 99.9% | 639.74 | 10 MG
SDP
Catalog No. 5000397250
Encompass_Preferred
Cyclic PSAP peptide is a cyclic pentapeptide with the sequence Cyclic(Asp-Trp-Leu-Pro-Lys), also shortened as Cyclic(DWLPK). It exhibits agent-like properties and has been shown to inhibit metastatic spread and restrain tumor development in vivo.
  • Cyclic pentapeptide
  • Sequence: Cyclic(Asp-Trp-Leu-Pro-Lys), shortened as Cyclic(DWLPK)
  • Exhibits agent-like properties
  • Inhibits metastatic spread
  • Restrains tumor development in vivo
Catalog No. 50-003-97250 Supplier Medchemexpress LLC Supplier No. HYP410210MG
Add to Cart
Add to Cart
Cyclic PSAP peptide is a cyclic pentapeptide with the sequence Cyclic(Asp-Trp-Leu-Pro-Lys), also shortened as Cyclic(DWLPK). It exhibits agent-like properties and has been shown to inhibit metastatic spread and restrain tumor development in vivo.
  • Cyclic pentapeptide
  • Sequence: Cyclic(Asp-Trp-Leu-Pro-Lys), shortened as Cyclic(DWLPK)
  • Exhibits agent-like properties
  • Inhibits metastatic spread
  • Restrains tumor development in vivo