Learn More
Medchemexpress LLC Cyclic somatostatin | 38916-34-6 | 99.9% | 1637.88 | 25 MG
SDP
Catalog No. 5000426821
Encompass_Preferred
Cyclic somatostatin (SRIF-14) is a growth hormone-release inhibiting factor that influences the contraction of ventricular cardiomyocytes. It is used in the research of severe, acute hemorrhages of gastroduodenal ulcers and cardiovascular disease. It appears as a white to off-white solid.
  • Inhibits contractile response of isoprenaline with an IC50 value of 13 nM
  • Neuropeptide co-stored with acetylcholine in cardiac parasympathetic innervation
  • Suitable for cardiovascular disease research
Catalog No. 50-004-26821 Supplier Medchemexpress LLC Supplier No. 25MGHYP0084
Cyclic somatostatin (SRIF-14) is a growth hormone-release inhibiting factor that influences the contraction of ventricular cardiomyocytes. It is used in the research of severe, acute hemorrhages of gastroduodenal ulcers and cardiovascular disease. It appears as a white to off-white solid.
  • Inhibits contractile response of isoprenaline with an IC50 value of 13 nM
  • Neuropeptide co-stored with acetylcholine in cardiac parasympathetic innervation
  • Suitable for cardiovascular disease research