Learn More
Medchemexpress LLC Cyclic somatostatin | 38916-34-6 | 99.9% | 1637.88 | 50 MG
SDP
Catalog No. 5000364410
Encompass_Preferred
Cyclic somatostatin (SRIF-14) is a growth hormone-release inhibiting factor primarily used in research for severe, acute hemorrhages of gastroduodenal ulcers. This neuropeptide, co-stored with acetylcholine in cardiac parasympathetic innervation, directly influences the contraction of ventricular cardiomyocytes and inhibits the contractile response of isoprenaline with an IC50 value of 13 nM.
  • Potent growth hormone-release inhibiting factor
  • Investigated for its role in severe, acute hemorrhages of gastroduodenal ulcers
  • Neuropeptide co-stored with acetylcholine in cardiac parasympathetic innervation
  • Directly affects the contraction of ventricular cardiomyocytes
  • Exhibits strong inhibitory effect on the contractile response of isoprenaline (IC50 value of 13 nM)
  • Valuable tool for cardiovascular disease research
Catalog No. 50-003-64410 Supplier Medchemexpress LLC Supplier No. 50MGHYP0084
Cyclic somatostatin (SRIF-14) is a growth hormone-release inhibiting factor primarily used in research for severe, acute hemorrhages of gastroduodenal ulcers. This neuropeptide, co-stored with acetylcholine in cardiac parasympathetic innervation, directly influences the contraction of ventricular cardiomyocytes and inhibits the contractile response of isoprenaline with an IC50 value of 13 nM.
  • Potent growth hormone-release inhibiting factor
  • Investigated for its role in severe, acute hemorrhages of gastroduodenal ulcers
  • Neuropeptide co-stored with acetylcholine in cardiac parasympathetic innervation
  • Directly affects the contraction of ventricular cardiomyocytes
  • Exhibits strong inhibitory effect on the contractile response of isoprenaline (IC50 value of 13 nM)
  • Valuable tool for cardiovascular disease research