Learn More
TARGETMOL CHEMICALS INC CYCLIC SOMATOSTATIN ACET 25MG
SDP
Catalog No. 502576168
Encompass_Preferred
Also available in 1 mg 10 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Cyclic somatostatin Acetate(38916-34-6(free base)) (Somatostatin-14) is a cyclic tetradecapeptide regulating many endocrine and nervous system functions. purity: 97%
Catalog No. 50-257-6168 Supplier TARGETMOL CHEMICALS INC Supplier No. TP1035L25MG
Also available in 1 mg 10 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Cyclic somatostatin Acetate(38916-34-6(free base)) (Somatostatin-14) is a cyclic tetradecapeptide regulating many endocrine and nervous system functions. purity: 97%