Learn More
Cayman Chemical Cyclic di-AMP ELISA Standard, 15 ng
SDP
Catalog No. 502942392
Encompass_Preferred
Cyclic di-AMP ELISA Standard has been tested and formulated to work exclusively with Cayman’s Cyclic di-AMP ELISA Kit
Catalog No. 50-294-2392 Supplier Cayman Chemical Supplier No. 40196415 NG
Cyclic di-AMP ELISA Standard has been tested and formulated to work exclusively with Cayman’s Cyclic di-AMP ELISA Kit