Learn More
Cell Signaling Technology Cyclin A2 (E6D1J) XP Rabbit Monoclonal Antibody, High Specificity, Validated for Multiple Applications, 100 µg
SDP
Supplier:  Cell Signaling Technology 67955S
Encompass
The Cyclin A2 (E6D1J) XP Rabbit Monoclonal Antibody is a highly specific and validated antibody for detecting human Cyclin A2 protein levels, playing a crucial role in cell cycle progression. Its rigorous validation for applications like Western Blotting, Immunoprecipitation, and Immunohistochemistry sets it apart, ensuring reliable results for scientific research.
Catalog No. NC2259524