Learn More
Enzo Life Sciences Cyclin B1 monoclonal antibody (V63) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC195E
Encompass_Preferred
Clone: V63. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the carboxy terminus of hamster cyclin B1. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Hamster, Guinea pig, Dog. Applications: IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7242