Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Cyclin C (E6V4Z) Rabbit Monoclonal Antibody, Human, Mouse, Unconjugated, 100 μl
SDP

Catalog No. 502732096
Encompass

The Cyclin C (E6V4Z) Rabbit Monoclonal Antibody is designed to detect endogenous levels of total cyclin C protein. Generated by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro263 of human cyclin C. Cyclin C functions as a component of the cyclin C/CDK8 complex, which associates with the Mediator Complex to regulate RNA polymerase II-dependent transcription, and plays significant roles in mitochondrial dynamics, apoptosis, and cell cycle progression.

  • Validated applications: western blotting, immunoprecipitation, immunofluorescence (immunocytochemistry), and flow cytometry (fixed/permeabilized).
  • Species reactivity: human and mouse.
  • Formulation: 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Research use only.
  • store at -20°C; do not aliquot the antibody.

Catalog No. 50-273-2096 Supplier Cell Signaling Technology Supplier No. 68179S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Cyclin C (E6V4Z) Rabbit Monoclonal Antibody is designed to detect endogenous levels of total cyclin C protein. Generated by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro263 of human cyclin C. Cyclin C functions as a component of the cyclin C/CDK8 complex, which associates with the Mediator Complex to regulate RNA polymerase II-dependent transcription, and plays significant roles in mitochondrial dynamics, apoptosis, and cell cycle progression.

  • Validated applications: western blotting, immunoprecipitation, immunofluorescence (immunocytochemistry), and flow cytometry (fixed/permeabilized).
  • Species reactivity: human and mouse.
  • Formulation: 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Research use only.
  • store at -20°C; do not aliquot the antibody.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.