Learn More
Diagnostic Biosystems CYCLIN D1 Cyclin D1 (EP12)
Catalog No. 502407458
Encompass_Preferred
Cyclin D1 belongs to the Cyclin D family. Cyclin D1 is required for the cell cycle G1/S transition. Amplification or overexpression of Cyclin D1 plays a pivotal role in the development of various human cancers including breast cancer, colon cancer, melanoma, prostate cancer and lymphoma. It is useful to differentiate mantle cell lymphoma from small cleaved cell lymphoma. Rabbit monoclonal antibodies to Cyclin D1 showed the highest sensitivity to detect this antigen in formalin fixed paraffin embedded tissue as compared to several other clones.
Catalog No. 50-240-7458 Supplier Diagnostic Biosystems Supplier No. RMPD106
Cyclin D1 belongs to the Cyclin D family. Cyclin D1 is required for the cell cycle G1/S transition. Amplification or overexpression of Cyclin D1 plays a pivotal role in the development of various human cancers including breast cancer, colon cancer, melanoma, prostate cancer and lymphoma. It is useful to differentiate mantle cell lymphoma from small cleaved cell lymphoma. Rabbit monoclonal antibodies to Cyclin D1 showed the highest sensitivity to detect this antigen in formalin fixed paraffin embedded tissue as compared to several other clones.