Learn More
Biomatik Corporation Cyclin D1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50160
Encompass_Preferred
Cyclin D1 Colorimetric Cell-Based ELISA Kit for detecting Cyclin D1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P24385. Gene ID: 595. Target Synonyms: BCL-1 oncogene; BCL1; cyclin D1; CYL-1; G1/S-specific cyclin D1; PRAD1; PRAD1 oncogene. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7417