Learn More
Enzo Life Sciences Cyclin D1 monoclonal antibody (5D4) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC200E
Encompass_Preferred
Clone: 5D4. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human cyclin D1. Species reactivity: Human, Mouse. Applications: Flow Cytometry, ICC, IHC (PS), IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7243