Learn More
Cell Signaling Technology Cell Signaling Technology Cyclin D1 (E3P5S) Rabbit Monoclonal Antibody, Human, Mouse, Rat, Monkey, 300 μl
SDP
Catalog No. NC3924510
Encompass
This rabbit monoclonal antibody recognizes endogenous levels of total cyclin D1 protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Ala284 of the human cyclin D1 protein to ensure high specificity and performance.
  • Reacts with human, mouse, rat, and monkey samples
  • Validated for western blotting, immunoprecipitation, immunohistochemistry (paraffin), immunofluorescence, and flow cytometry
  • Supplied in a buffer containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 µg/ml BSA, 50% glycerol, and less than 0.02% sodium azide
  • Demonstrates a molecular weight of 36 kDa
  • For research use only
Catalog No. NC3924510 Supplier Cell Signaling Technology Supplier No. 55506L
This rabbit monoclonal antibody recognizes endogenous levels of total cyclin D1 protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Ala284 of the human cyclin D1 protein to ensure high specificity and performance.
  • Reacts with human, mouse, rat, and monkey samples
  • Validated for western blotting, immunoprecipitation, immunohistochemistry (paraffin), immunofluorescence, and flow cytometry
  • Supplied in a buffer containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 µg/ml BSA, 50% glycerol, and less than 0.02% sodium azide
  • Demonstrates a molecular weight of 36 kDa
  • For research use only