Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclin D2/CCND2 antibody (YA789) | Affinity purified | Predicted band size: 33 kDa; Observed band size: 38 kDa | 10 UL
SDP

Catalog No. 5000396037
Encompass_Preferred

Cyclin D2/CCND2 antibody (YA789) is a mouse-derived, non-conjugated IgG2b monoclonal antibody targeting Cyclin D2/CCND2. It is formulated in 1x PBS (pH 7.3), 50% glycerol, and 0.5% BSA, containing 0.02% sodium azide as a preservative. This antibody is intended for research use only and reacts with human species.

  • Host: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG2b
  • Species reactivity: Human
  • Purity: Affinity purified
  • Applications: Western Blot (WB)
  • Immunogen: Synthetic peptide corresponding to human Cyclin D2
  • Storage: Store at -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-003-96037 Supplier Medchemexpress LLC Supplier No. HYP8063410UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclin D2/CCND2 antibody (YA789) is a mouse-derived, non-conjugated IgG2b monoclonal antibody targeting Cyclin D2/CCND2. It is formulated in 1x PBS (pH 7.3), 50% glycerol, and 0.5% BSA, containing 0.02% sodium azide as a preservative. This antibody is intended for research use only and reacts with human species.

  • Host: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG2b
  • Species reactivity: Human
  • Purity: Affinity purified
  • Applications: Western Blot (WB)
  • Immunogen: Synthetic peptide corresponding to human Cyclin D2
  • Storage: Store at -20°C for 1 year, avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.