Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclin D3/CCND3 Antibody (YA484) | 33 kDa | 100 UL
SDP

Catalog No. 5000397714
Encompass_Preferred

Cyclin D3/CCND3 Antibody (YA484) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cyclin D3/CCND3. This protein is a regulatory component of the cyclin D3-CDK4 complex, crucial for regulating the cell cycle during G(1)/S transition by phosphorylating retinoblastoma (RB) protein family members.

  • Host: Rabbit
  • Reactivity: Human, Mouse
  • Clonality: Monoclonal
  • Target: Cyclin D3/CCND3
  • Applications: WB, ICC/IF
  • Storage: Store at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Formulation: Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol. Preservative: 0.05% Sodium Azide.
  • Purity: Protein A affinity purified.

Catalog No. 50-003-97714 Supplier Medchemexpress LLC Supplier No. HYP80099100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclin D3/CCND3 Antibody (YA484) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cyclin D3/CCND3. This protein is a regulatory component of the cyclin D3-CDK4 complex, crucial for regulating the cell cycle during G(1)/S transition by phosphorylating retinoblastoma (RB) protein family members.

  • Host: Rabbit
  • Reactivity: Human, Mouse
  • Clonality: Monoclonal
  • Target: Cyclin D3/CCND3
  • Applications: WB, ICC/IF
  • Storage: Store at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Formulation: Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol. Preservative: 0.05% Sodium Azide.
  • Purity: Protein A affinity purified.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.