Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclin D3/CCND3 Antibody (YA484) | Protein A affinity purified | 33 kDa | 10 UL
SDP

Catalog No. 5000397520
Encompass_Preferred

Cyclin D3/CCND3 Antibody (YA484) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Cyclin D3/CCND3. This antibody is supplied in 1*TBS (pH 7.4) containing 0.05% BSA and 40% Glycerol, with 0.05% Sodium Azide as a preservative. It is suitable for applications such as Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF).

  • Host: Rabbit
  • Reactivity: Human, Mouse
  • Clonality: Monoclonal antibody
  • Purity: Protein A affinity purified
  • Formulation: 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, 0.05% Sodium Azide
  • Storage: Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Application: WB (1:500-1:1000)
  • Application: ICC/IF (1:50-1:100)
  • Background: Functions as a regulatory component of the cyclin D3-CDK4 complex, regulating cell cycle during G(1)/S transition by phosphorylating and inhibiting retinoblastoma protein family members.

Catalog No. 50-003-97520 Supplier Medchemexpress LLC Supplier No. HYP8009910UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclin D3/CCND3 Antibody (YA484) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Cyclin D3/CCND3. This antibody is supplied in 1*TBS (pH 7.4) containing 0.05% BSA and 40% Glycerol, with 0.05% Sodium Azide as a preservative. It is suitable for applications such as Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF).

  • Host: Rabbit
  • Reactivity: Human, Mouse
  • Clonality: Monoclonal antibody
  • Purity: Protein A affinity purified
  • Formulation: 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, 0.05% Sodium Azide
  • Storage: Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Application: WB (1:500-1:1000)
  • Application: ICC/IF (1:50-1:100)
  • Background: Functions as a regulatory component of the cyclin D3-CDK4 complex, regulating cell cycle during G(1)/S transition by phosphorylating and inhibiting retinoblastoma protein family members.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.