Learn More
OriGene Cyclin E1 (CCNE1) (NM_001238) Human Tagged ORF Clone Lentiviral Particle 200 ul
SDP
Catalog No. NC3351929
Encompass_Preferred
Lenti ORF particles, CCNE1 (mGFP-tagged) - Human cyclin E1 (CCNE1), transcript variant 1, 200ul, >10^7 TU/mL
Catalog No. NC3351929 Supplier OriGene Supplier No. RC204289L4V
Lenti ORF particles, CCNE1 (mGFP-tagged) - Human cyclin E1 (CCNE1), transcript variant 1, 200ul, >10^7 TU/mL