Learn More
Sino Biological Recombinant Human cyclin E2 Protein (Full Length, GST Tag)
Catalog No. 502575447
Encompass_Preferred
Recombinant full length human CyclinE2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5447 Supplier Sino Biological Supplier No. C8230BG100
Recombinant full length human CyclinE2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.