Learn More
Sino Biological Recombinant Human Cyclin D3/CCND3 Protein (Full Length, GST Tag)
Catalog No. 502575452
Encompass_Preferred
Recombinant full length human CyclinD3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5452 Supplier Sino Biological Supplier No. C8130B3G1000
Recombinant full length human CyclinD3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.