Learn More
Apexbio Technology LLC Cyclizine 2HCl 5897-18-7 50mg
SDP
Catalog No. 5000092050
Encompass_Preferred
Cyclizine 2HCl (CAS 5897-18-7) is a synthetic piperazine derivative that functions as an antagonist at histamine H1 receptors By inhibiting H1 receptor-mediated signaling Cyclizine 2HCl effectively suppresses histamine-induced physiological responses including those involved in allergic reactions and motion sickness This compound is widely utilized in biomedical research to investigate histaminergic pathways allergic mechanisms and the pharmacological modulation of H1 receptor activity Its antagonist properties make it valuable for studies examining the mechanistic basis of histamine-mediated processes and for evaluating novel anti-allergic therapeutics
Catalog No. 50-000-92050 Supplier Apexbio Technology LLC Supplier No. B154850
Cyclizine 2HCl (CAS 5897-18-7) is a synthetic piperazine derivative that functions as an antagonist at histamine H1 receptors By inhibiting H1 receptor-mediated signaling Cyclizine 2HCl effectively suppresses histamine-induced physiological responses including those involved in allergic reactions and motion sickness This compound is widely utilized in biomedical research to investigate histaminergic pathways allergic mechanisms and the pharmacological modulation of H1 receptor activity Its antagonist properties make it valuable for studies examining the mechanistic basis of histamine-mediated processes and for evaluating novel anti-allergic therapeutics