Learn More
Selleck Chemical LLC Cyclizine 2HCl S4139-50mg
SDP
Catalog No. 503108670
Encompass_Preferred
Cyclizine 2HCl is a piperazine derivative with Histamine H1 receptor antagonist activity
Catalog No. 50-310-8670 Supplier Selleck Chemical LLC Supplier No. S413950MG
Cyclizine 2HCl is a piperazine derivative with Histamine H1 receptor antagonist activity