Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cyclizine hydrochloride, 98%

Catalog No. AC461040010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461040010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC461040010 Supplier Thermo Scientific Chemicals Supplier No. 461040010
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 303-25-3 | C18H22N2·HCl | 302.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-25-3
Molecular Formula C18H22N2·HCl
Molecular Weight (g/mol) 302.84
InChI Key UKPBEPCQTDRZSE-UHFFFAOYSA-N
Synonym cyclizine hydrochloride, cyclizine hcl, dihydrochlorid, marzine, reis-fit, unii-w0o1nhp4we, marezine hydrochloride, cyclizine monohydrochloride, w0o1nhp4we, 1-diphenylmethyl-4-methylpiperazine monohydrochloride
PubChem CID 6420019
ChEBI CHEBI:51045
IUPAC Name 1-benzhydryl-4-methylpiperazine;hydrochloride
SMILES CN1CCN(CC1)C(C2=CC=CC=C2)C3=CC=CC=C3.Cl

Specifications

Melting Point 291.0°C to 293.0°C
Loss on Drying 1% max. (120°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 302.84
Percent Purity 98%
Chemical Name or Material Cyclizine hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/fa
EINECSNumber 206-136-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.