Learn More
CPC Scientific Cyclo(-Arg-Gly-Asp-D-Tyr-Lys) (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110381
Encompass
alphav 3 Integrin Binding RGD Peptide RGD Tumor Targeting and Tumor Imaging PeptideSEQUENCE: Cyclo(-Arg-Gly-Asp-D-Tyr-Lys) (trifluoroacetate salt)ONE-LETTER SEQUENCE: Cyclo(-RGDyK)MOLECULAR FORMULA: C27H41N9O8MOLECULAR WEIGHT: 619.68STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [217099-14-4]SYNONYMS: Cyclo(-Arg-Gly-Asp-D-Tyr-Lys)RESEARCH AREA: CancerREFERENCES:R. Haubner, et al., J. Nuclear Med., 42, 326 (2001).X. Chen, et al., Nucl. Med. Biol., 31, 179 (2004).X. Chen, et al., Cancer Res., 641, 8009 (2004).SKU(s): RGDP-005A, RGDP-005B
Catalog No. 50-211-0381 Supplier CPC Scientific Supplier No. RGDP005A
alphav 3 Integrin Binding RGD Peptide RGD Tumor Targeting and Tumor Imaging PeptideSEQUENCE: Cyclo(-Arg-Gly-Asp-D-Tyr-Lys) (trifluoroacetate salt)ONE-LETTER SEQUENCE: Cyclo(-RGDyK)MOLECULAR FORMULA: C27H41N9O8MOLECULAR WEIGHT: 619.68STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [217099-14-4]SYNONYMS: Cyclo(-Arg-Gly-Asp-D-Tyr-Lys)RESEARCH AREA: CancerREFERENCES:R. Haubner, et al., J. Nuclear Med., 42, 326 (2001).X. Chen, et al., Nucl. Med. Biol., 31, 179 (2004).X. Chen, et al., Cancer Res., 641, 8009 (2004).SKU(s): RGDP-005A, RGDP-005B