Learn More
Selleck Chemical LLC Cyclo
Supplier: Selleck Chemical LLC S78441G
Cyclo(RGDyK) TFA is a salt formed by the combination of Cyclo(RGDyK) and trifluoroacetate (TFA) It is an effective and selective Integrin v 3 inhibitor with a low IC50 value (e g 20 nM) Cyclo(RGDyK) TFA is of great significance in the treatment of cardiovascular diseases osteoporosis inflammation and tumors