Learn More
Selleck Chemical LLC Cyclo
SDP
Catalog No. 5000001635
Encompass_Preferred
Cyclo(RGDyK) TFA is a salt formed by the combination of Cyclo(RGDyK) and trifluoroacetate (TFA) It is an effective and selective Integrin v 3 inhibitor with a low IC50 value (e g 20 nM) Cyclo(RGDyK) TFA is of great significance in the treatment of cardiovascular diseases osteoporosis inflammation and tumors
Catalog No. 50-000-01635 Supplier Selleck Chemical LLC Supplier No. S784410MM/1ML
Cyclo(RGDyK) TFA is a salt formed by the combination of Cyclo(RGDyK) and trifluoroacetate (TFA) It is an effective and selective Integrin v 3 inhibitor with a low IC50 value (e g 20 nM) Cyclo(RGDyK) TFA is of great significance in the treatment of cardiovascular diseases osteoporosis inflammation and tumors