Learn More
Selleck Chemical LLC Cyclo
SDP
Catalog No. 5000003609
Encompass_Preferred
Cyclo(-RGDfK) TFA is the salt product of Cyclo(-RGDfK) and trifluoroacetic acid (TFA) The addition of TFA enhances stability and biocompatibility Cyclo(-RGDfK) TFA effectively targets tumor microvessels and cancer cells by specifically binding to the cell surface Integrin v 3
Catalog No. 50-000-03609 Supplier Selleck Chemical LLC Supplier No. S783425MG
Add to Cart
Add to Cart
Cyclo(-RGDfK) TFA is the salt product of Cyclo(-RGDfK) and trifluoroacetic acid (TFA) The addition of TFA enhances stability and biocompatibility Cyclo(-RGDfK) TFA effectively targets tumor microvessels and cancer cells by specifically binding to the cell surface Integrin v 3