Learn More
Cayman Chemical Cyclo(Δ-Ala-L-Val), Purity: ≥98%, CAS: 25516-00-1, 5 mg
SDP
Catalog No. 502932828
Encompass_Preferred
A bacterial cyclic dipeptide; activates a LuxR-based AHL E. coli biosensor and blocks activation of the biosensor by the quorum sensing signal molecule 3-oxo-C6-HSL (IC50 = 0.8 mM); reduces S. liquefaciens colony expansion by 21% at 15 μM; inhibits interaction of kinases Ras and Raf-1 in a yeast two-hybrid assay in a concentration-dependent manner
Catalog No. 50-293-2828 Supplier Cayman Chemical Supplier No. 257395 MG
A bacterial cyclic dipeptide; activates a LuxR-based AHL E. coli biosensor and blocks activation of the biosensor by the quorum sensing signal molecule 3-oxo-C6-HSL (IC50 = 0.8 mM); reduces S. liquefaciens colony expansion by 21% at 15 μM; inhibits interaction of kinases Ras and Raf-1 in a yeast two-hybrid assay in a concentration-dependent manner