Learn More
Bachem Cyclo(-Arg-Gly-Asp-D-Phe-Cys)
SDP
Supplier:  Bachem 4069272.0001
Encompass_Preferred
Cyclo(-Arg-Gly-Asp-D-Phe-Cys) Acetate, 862772-11-0, C24H34N8O7S, Mr 578.65, 1mg. c(RGDfC), avb3 Integrin Binding Cyclic RGD Peptide. c(RGDfC), a RGD tumor-targeting peptide, which binds to avb3 receptors. c(RGDfC) can be easily conjugated to drugs as combretastatin or surfaces.
Catalog No. 50-383-345