Learn More
Medchemexpress LLC Cyclo(Arg-Gly-Asp-D-Phe-Cys) | 862772-11-0 | 98.1% | 578.64 | 1 MG
SDP
Catalog No. 5000424220
Encompass_Preferred
Cyclo(Arg-Gly-Asp-D-Phe-Cys) (Cyclo RGDfC) is a cyclic RGD peptide with high affinity to αvβ3 that disrupts cell integrin interactions. It inhibits pluripotent marker expression in embryonic stem cells (ESCs) and the tumorigenic potential of mESCs in vivo. This product is for research use only and can be utilized in tumor research.
  • High affinity to αvβ3
  • Disrupts cell integrin interactions
  • Inhibits pluripotent marker expression in embryonic stem cells (ESCs)
  • Inhibits tumorigenic potential of mESCs in vivo
  • Used in tumor research
  • Solid, white to off-white appearance
Catalog No. 50-004-24220 Supplier Medchemexpress LLC Supplier No. HYP23001MG
Cyclo(Arg-Gly-Asp-D-Phe-Cys) (Cyclo RGDfC) is a cyclic RGD peptide with high affinity to αvβ3 that disrupts cell integrin interactions. It inhibits pluripotent marker expression in embryonic stem cells (ESCs) and the tumorigenic potential of mESCs in vivo. This product is for research use only and can be utilized in tumor research.
  • High affinity to αvβ3
  • Disrupts cell integrin interactions
  • Inhibits pluripotent marker expression in embryonic stem cells (ESCs)
  • Inhibits tumorigenic potential of mESCs in vivo
  • Used in tumor research
  • Solid, white to off-white appearance