Learn More
Medchemexpress LLC Cyclo(Arg-Gly-Asp-D-Phe-Cys) TFA | 98.7% | 692.66 | 10 MG
SDP
Catalog No. 5000424441
Encompass_Preferred
Cyclo(Arg-Gly-Asp-D-Phe-Cys) (Cyclo RGDfC) TFA is a cyclic RGD peptide known for its high affinity to αvβ3, allowing it to disrupt cell integrin interactions. This compound is capable of inhibiting pluripotent marker expression in embryonic stem cells (ESCs) and reducing the tumorigenic potential of mESCs in vivo. It is relevant for research concerning tumors.
  • High affinity to αvβ3.
  • Disrupts cell integrin interactions.
  • Inhibits pluripotent marker expression in embryonic stem cells (ESCs).
  • Reduces the tumorigenic potential of mESCs in vivo.
  • Applicable in tumor research.
Catalog No. 50-004-24441 Supplier Medchemexpress LLC Supplier No. HYP2300A10MG
Cyclo(Arg-Gly-Asp-D-Phe-Cys) (Cyclo RGDfC) TFA is a cyclic RGD peptide known for its high affinity to αvβ3, allowing it to disrupt cell integrin interactions. This compound is capable of inhibiting pluripotent marker expression in embryonic stem cells (ESCs) and reducing the tumorigenic potential of mESCs in vivo. It is relevant for research concerning tumors.
  • High affinity to αvβ3.
  • Disrupts cell integrin interactions.
  • Inhibits pluripotent marker expression in embryonic stem cells (ESCs).
  • Reduces the tumorigenic potential of mESCs in vivo.
  • Applicable in tumor research.