Learn More
Medchemexpress LLC Cyclo(Arg-Gly-Asp-(D-Phe)-Glu) (c(RGDfE)) | 756500-22-8 | 99.9% | 604.61 | 50 MG
SDP
Catalog No. 5000398218
Encompass_Preferred
Cyclo(Arg-Gly-Asp-(D-Phe)-Glu), also known as c(RGDfE), is a cyclic RGD peptide designed to target integrin αvβ3. It is utilized in various research applications, including the modification of drug-loaded nanoparticles and in studies related to cancer, particularly pancreatic cancer.
  • Targets integrin αvβ3.
  • Used for modifying drug-loaded nanoparticles.
  • Employed in cancer research, especially pancreatic cancer.
  • Enhances targeting of nanoparticles towards BxPC3 cells and integrin αvβ3 expressing cells in vitro.
  • Enhances targeting of nanoparticles to BxPC3 tumors in vivo.
Catalog No. 50-003-98218 Supplier Medchemexpress LLC Supplier No. HYP502150MG
Cyclo(Arg-Gly-Asp-(D-Phe)-Glu), also known as c(RGDfE), is a cyclic RGD peptide designed to target integrin αvβ3. It is utilized in various research applications, including the modification of drug-loaded nanoparticles and in studies related to cancer, particularly pancreatic cancer.
  • Targets integrin αvβ3.
  • Used for modifying drug-loaded nanoparticles.
  • Employed in cancer research, especially pancreatic cancer.
  • Enhances targeting of nanoparticles towards BxPC3 cells and integrin αvβ3 expressing cells in vitro.
  • Enhances targeting of nanoparticles to BxPC3 tumors in vivo.