Learn More
Carbosynth LLC. Cyclo(Arg-Gly-Asp-D-Phe-Lys), 1mg, MW: (M.W. 603.68), Molecular formula: C27H41N9O7, CAS# [161552-03-0]
SDP
Supplier:  Carbosynth LLC. PCI3661PI1MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: cyclo (Arg-Gly-Asp-D-Phe-Lys), c(RGDfK), Application: v3 Integrin Binding RGD Peptide RGD Tumor Targeting Peptide(Requires further derivatization before use), Salt Form:, Acetate salt, Storage Temperature: -20C, References: M. Kantlehner, P. Schaffner, D. Finsinger, J. Meyer, A. Jonczyk, B. Diefenbach, B. Nies, G. Hozemann, S.L. Goodman, and H. Kessler. Chembiochem., 1, 107 (2000), R.J. Kok, A.J. Schraa, E.J. Bos, H.E. Moorlag, S.A. Ásgeirsdóttir, M. Everts, DK. Meijer, and G. Molema, Bioconjug. Chem., 13, 128 (2002).
Catalog No. 50-168-6273