Learn More
Carbosynth LLC. Cyclo(Arg-Gly-Asp-D-Tyr-Lys), 5mg, MW: (M.W. 619.68), Molecular formula: C27H41N9O8
SDP
Catalog No. 501686278
Encompass_Preferred
Synonyms: cyclo (Arg-Gly-Asp-D-Tyr-Lys), c(RGDyK), Application: alphavbeta3 Integrin Binding RGD Peptide RGD Tumor Targeting and Tumor Imaging Peptide (Requires further derivatization before use), Storage Temperature: -20°C, References: R. Haubner, H.J. Wester, F. Burkhart, R. Senekowitsch-Schmidtky, W. Weber, S.L. Goodman, H. Kessler, and M. Schwaiger, J. NuCLEAR™ Med., 42, 326 (2001), X. Chen, R. Park. A.H. Shahinian, M. Tohme, V. Khankaldyyan, M.H. Bozorgzdeh, J.R. Bading, R. Moats, W.E. Laug, and P.S. Conti, Nucl. Med. Biol., 31, 179 (2004)., X. Chen, P.S. Conti, and R.A. Moats, Cancer Res., 641, 8009 (2004).
Catalog No. 50-168-6278 Supplier Carbosynth LLC. Supplier No. PCI3662PI5MG
Add to Cart
Add to Cart
Synonyms: cyclo (Arg-Gly-Asp-D-Tyr-Lys), c(RGDyK), Application: alphavbeta3 Integrin Binding RGD Peptide RGD Tumor Targeting and Tumor Imaging Peptide (Requires further derivatization before use), Storage Temperature: -20°C, References: R. Haubner, H.J. Wester, F. Burkhart, R. Senekowitsch-Schmidtky, W. Weber, S.L. Goodman, H. Kessler, and M. Schwaiger, J. NuCLEAR™ Med., 42, 326 (2001), X. Chen, R. Park. A.H. Shahinian, M. Tohme, V. Khankaldyyan, M.H. Bozorgzdeh, J.R. Bading, R. Moats, W.E. Laug, and P.S. Conti, Nucl. Med. Biol., 31, 179 (2004)., X. Chen, P.S. Conti, and R.A. Moats, Cancer Res., 641, 8009 (2004).